Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAD2L1BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565029 Supplier Invitrogen™ Supplier No. PA565029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDLEWYEKSEE THASQIELLE TSSTQEPLNA SEAFCPRDCM VPVVFPGPVS QEGCCQFTCE LLKHIMYQRQ QLPLPY Highest antigen sequence identity to the following orthologs - mouse 72%, rat 73%.

The protein encoded by this gene was identified as a binding protein of the MAD2 mitotic arrest deficient-like 1 (MAD2/MAD2L1). MAD2 is a key component of the spindle checkpoint that delays the onset of anaphase until all the kinetochores are attached to the spindle. This protein may interact with the spindle checkpoint and coordinate cell cycle events in late mitosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD2L1BP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAD2L1BP
Gene Accession No. Q15013
Gene Alias 0610009D16Rik; 2510031P20Rik; AU018945; AW550977; C76045; caught by MAD2 protein; CMT2; CMT2A; CMT2B; KIAA0110; MAD2L1 binding protein; MAD2L1-binding protein; Mad2l1bp; Mad2lbp; mKIAA0110; p31(comet); RP1-261G23.6
Gene Symbols MAD2L1BP
Host Species Rabbit
Immunogen Recombinant Human MAD2L1BP. Recombinant protein control fragment (Product #RP-104592).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.