Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAD2L1BP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575530
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene was identified as a binding protein of the MAD2 mitotic arrest deficient-like 1 (MAD2/MAD2L1). MAD2 is a key component of the spindle checkpoint that delays the onset of anaphase until all the kinetochores are attached to the spindle. This protein may interact with the spindle checkpoint and coordinate cell cycle events in late mitosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
MAD2L1BP | |
Polyclonal | |
Unconjugated | |
MAD2L1BP | |
0610009D16Rik; 2510031P20Rik; AU018945; AW550977; C76045; caught by MAD2 protein; CMT2; CMT2A; CMT2B; KIAA0110; MAD2L1 binding protein; MAD2L1-binding protein; Mad2l1bp; Mad2lbp; mKIAA0110; p31(comet); RP1-261G23.6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
316237, 66591, 9587 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15013, Q9DCX1 | |
MAD2L1BP | |
Synthetic peptide corresponding to amino acids 40-85 of Human MAD2L1BP. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction