Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD2L1BP Recombinant Rabbit Monoclonal Antibody (7W8H1)
GREENER_CHOICE

Catalog No. PIMA542712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542712 Supplier Thermo Scientific Supplier No. MA542712
Add to Cart
Add to Cart
This item is not returnable. View return policy

MAD2L1BP Recombinant Rabbit Monoclonal Antibody (7W8H1)

MAD2L1BP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD2L1BP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 7W8H1
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAD2L1BP
Gene Accession No. Q15013, Q9DCX1
Gene Alias 0610009D16Rik; 2510031P20Rik; AU018945; AW550977; C76045; caught by MAD2 protein; CMT2; CMT2A; CMT2B; KIAA0110; MAD2L1 binding protein; MAD2L1-binding protein; Mad2l1bp; Mad2lbp; mKIAA0110; p31(comet); RP1-261G23.6
Gene Symbols MAD2L1BP
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 175-274 of human MAD2L1BP (Q15013).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316237, 66591, 9587
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.