Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAD2L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120177 Supplier Invitrogen™ Supplier No. PA5120177
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SH-SY5Y, A375, K-562, SW480, HeLa, Mouse liver, Rat liver. The target is usually found in the following locations: Cytoplasm, Nucleus, cytoskeleton, spindle. Immunogen sequence: MTTLTRQDLN FGQVVADVLC EFLEVAVHLI LYVREVYPVG IFQKRKKYNV PVQMSCHPEL NQYIQDTLHC VKPLLEKNDV EKVVVVILDK EHRPVEKFVF EITQPPLLSI SSDSLLSHVE QLLRAFILKI SVCDAVLDHN PPGCTFTVLV HTREAATRNM EKIQVIKDFP WILADEQDVH MHDPRLIPLK TMTSDILKMQ LYVEERAHKG S.

The protein encoded by this gene is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. The encoded protein, which is similar to MAD2L1, is capable of interacting with ADAM9, ADAM15, REV1, and REV3 proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD2L2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAD2L2
Gene Accession No. D3Z8D9, Q9D752, Q9UI95
Gene Alias 2310033C13Rik; G1-453-4; hREV7; MAD2 (mitotic arrest deficient, yeast, homolog)-like 2; MAD2 mitotic arrest deficient-like 2; MAD2 mitotic arrest deficient-like 2 (yeast); Mad2b; Mad2l2; MAD2-like protein 2; Mitotic arrest deficient 2-like protein 2; mitotic arrest deficient homolog-like 2; mitotic spindle assembly checkpoint protein MAD2B; polymerase (DNA-directed), zeta 2, accessory subunit; POLZ2; repro22; REV7; REV7 homolog; RP3-330O12.4
Gene Symbols MAD2L2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-211 of human MAD2B/MAD2B/MAD2L2 (NP_0063323).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10459, 313702, 71890
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.