Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAD4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001117
Description
MAD4 Polyclonal specifically detects MAD4 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
MAD4 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
BHLHC12, bHLHc12Max-interacting transcriptional repressor MAD4, Class C basic helix-loop-helix protein 12, Mad4 homolog, MAD4Max-associated protein 4, MAX dimerization protein 4, Max dimerizer 4, MST149, MSTP149, MXD4 MAX dimerization protein 4 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
The appearance of the 22 kDa band was blocked by incubation with the immunising peptide. Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.5 - 2 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_006445. | |
MXD4 | |
Peptide with sequence GPHCRRLGRPALS corresponding to C-Terminus according to NP_006445.1. | |
0.1 mg | |
Cell Cycle and Replication | |
10608 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction