Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119837 Supplier Invitrogen™ Supplier No. PA5119837
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Mouse brain tissue lysate, mouse lung tissue lysate. Subcellular Location: Nucleus.

This gene is a member of the MAD gene family. The MAD genes encode basic helix-loop-helix-leucine zipper proteins that heterodimerize with MAX protein, forming a transcriptional repression complex. The MAD proteins compete for MAX binding with MYC, which heterodimerizes with MAX forming a transcriptional activation complex. Studies in rodents suggest that the MAD genes are tumor suppressors and contribute to the regulation of cell growth in differentiating tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene MXD4
Gene Accession No. Q14582, Q60948
Gene Alias 2810410A03Rik; bHLHc12; Class C basic helix-loop-helix protein 12; LOW QUALITY PROTEIN: max dimerization protein 4; MAD4; Mad4 homolog; max dimerization protein 4; Max dimerizer 4; max-associated protein 4; Max-interacting transcriptional repressor MAD4; MST149; MSTP149; MXD4
Gene Symbols MXD4
Host Species Rabbit
Immunogen Recombinant protein within human MAD4 aa 1-180.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10608, 17122
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.