Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAdCAM-1 Antibody (683715) [Alexa Fluor™ 532], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6056X
Description
MAdCAM-1 Monoclonal antibody specifically detects MAdCAM-1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
MAdCAM-1 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8174 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, CyTOF | |
683715 | |
Flow Cytometry, CyTOF-ready | |
hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
Mouse myeloma cell line NS0-derived recombinant human MAdCAM-1, Leu21-Gln333, Accession # AAY82472 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction