Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAdCAM-1 Antibody (OTI1A5), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272568AF532
Description
MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MAdCAM-1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
MADCAM1 | |
Full length recombinant protein of human MAdCAM-1 (NP_570116) produced in HEK293T cells. | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1A5 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
Mouse | |
0.1 mL | |
8174 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction