Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAdCAM-1 Antibody (RM0284-11A55), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211785AF488
Description
MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot.Specifications
MAdCAM-1 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MADCAM1 | |
Mouse recombinant MAdCAM-1 (extracellular domain) | |
0.1 mL | |
8174 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0284-11A55 | |
Western Blot | |
hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction