Learn More
MAdCAM-1 Rat anti-Mouse, Brilliant Violet 510, Clone: MECA-89, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 744107
Description
The MECA-89 antibody specifically recognizes the mucosal vascular addressin cell adhesion molecule (MAdCAM-1). In the fetus and neonate, MAdCAM-1 is the predominant vascular addressin on the high endothelial venules (HEV) of peripheral lymph nodes. In adult mice, MAdCAM-1 is preferentially expressed in mucosal lymphoid tissues and lamina propria; it is also expressed on sinus-lining cells in the spleen. MAdCAM-1 expression is upregulated on the HEV of peripheral lymph nodes in adult NOD mice and is involved in the development of diabetes and insulitis. Furthermore, there is evidence that IFN-γcan induce MAdCAM-1 expression in non-mucosal sites in adult mice. MAdCAM-1 is a predominant ligand for integrinα4β7, a lymphocyte mucosal homing receptor, and a facultative ligand for CD62L (L-selectin). MECA-89 mAb binds to the second domain of MAdCAM-1 and does not block MAdCAM-1-dependent binding in vitro /. Source of the immunogen was endothelial cells from BALB/c mouse mesenteric and peripheral lymph nodes.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
MAdCAM-1 | |
MECA-89 | |
Brilliant Violet 510 | |
MAdCAM-1, Madcam1, mMAdCAM-1, MADCA | |
Mouse mesenteric and peripheral lymph node cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.