Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAdCAM-1 Rat anti-Mouse, Brilliant Violet 786, Clone: MECA-367, BD Optibuild™

Catalog No. BDB742817 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742817 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742817 Supplier BD Biosciences Supplier No. 742817
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MECA-367 monoclonal antibody specifically recognizes mucosal vascular addressin MAdCAM-1. In the fetus and neonate, MAdCAM-1 is the predominant vascular addressin on the high endothelial venules (HEV) of peripheral lymph nodes. In adult mice, MAdCAM-1 is preferentially expressed in mucosal lymphoid tissues and lamina propria; it is also expressed on sinus-lining cells in the spleen. MAdCAM-1 expression is upregulated on the HEV of peripheral lymph nodes in adult NOD mice4 and is involved in the development of diabetes and insulitis. Furthermore, there is evidence that IFN-γcan induce MAdCAM-1 expression in non-mucosal sites in adult mice. MAdCAM-1 is a predominant ligand for integrinα4β7, a lymphocyte mucosal homing receptor, and a facultative ligand for CD62L (L-selectin). MECA-367 mAb binds to the first domain of MAdCAM-1 and blocks MAdCAM-1-dependent binding in vitro /and lymphocyte homing to Peyer's patch HEV in vivo /. Source of the immunogen was endothelial cells from BALB/c mouse mesenteric and peripheral lymph nodes.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAdCAM-1
Classification Monoclonal
Clone MECA-367
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Madcam1, MADCA, mucosal vascular addressin cell adhesion molecule 1
Host Species Rat
Immunogen Mouse endothelial cells from BALB/c mouse mesenteric and peripheral lymph nodes.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.