Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAdCAM-1 Rat anti-Mouse, Unconjugated, Clone: MECA-367, MilliporeSigma™
Rat Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | MAdCAM-1 |
---|---|
Clone | MECA-367 |
Concentration | Please refer to lot specific datasheet. |
Applications | Immunohistochemistry (Paraffin), Inhibition Assays, Neutralization, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF2072-25
|
MilliporeSigma
MABF207225UG |
25 μg |
Each for $192.35
|
|
|||||
MABF2072-100
|
MilliporeSigma
MABF2072100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-MAdCAM-1, clone MECA-367, Cat. No. MABF2072, is a rat monoclonal antibody that detects murine Mucosal addressin cell adhesion molecule 1 (MAdCAM-1) and has been tested for use in Immunohistochemistry (Paraffin), Inhibition/Function assays, Neutralizing, and Western Blotting.Specifications
MAdCAM-1 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
Mucosal addressin cell adhesion molecule 1;mMAdCAM-1 | |
Madcam1 | |
IgG2a κ | |
Protein G purified | |
Clone MECA-367 detects Mucosal addressin cell adhesion molecule 1 (MAdCAM-1) in murine cells. | |
43.65 kDa calculated. |
MECA-367 | |
Immunohistochemistry (Paraffin), Inhibition Assays, Neutralization, Western Blot | |
Unconjugated | |
Rat | |
Inflammation & Immunology | |
Purified rat monoclonal antibody IgG2a in PBS without azide. | |
XP_006513356.1 | |
Endothelial cells isolated from BALB/c mouse mesenteric and peripheral lymph nodes. | |
Primary | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title