Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAdCAM1 Monoclonal Antibody (CA102.2C1), eBioscience™
GREENER_CHOICE

Catalog No. 5017839
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-178-39 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-178-39 Supplier Invitrogen™ Supplier No. BMS170
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This antigen is expressed in mucosal lymphoid tissue and lamina propria and is a counter-receptor for CD62L and CD49d. MAdCAM-1 is involved in homing of subpopulation of memory T cells to the gut lamina propria, and recirculation of naive lymphocytes in Peyer's patches and mesenteric lymph nodes. The 1F3F3D4 antibody reacts with mouse tumor necrosis factor-alpha (TNF alpha), a 17 kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4(+) T cells. Applications Tested: ELISA, Functional Studies (Blocking), Western Blotting.

The protein encoded by this gene is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAdCAM1
Applications ELISA, Functional Assay, Western Blot
Classification Monoclonal
Clone CA102.2C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MADCAM1
Gene Accession No. Q13477
Gene Alias AV211525; hMAdCAM-1; MACAM1; MADCAM1; MAdCAM-1; mMAdCAM-1; mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1; mucosal vascular addressin cell adhesion molecule 1; rMAdCAM-1
Gene Symbols MADCAM1
Host Species Mouse
Immunogen Rec. Human MadCAM-1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.