Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAdCAM1 Monoclonal Antibody (MECA-367), Biotin, eBioscience™
GREENER_CHOICE

Catalog No. 5012122
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-121-22 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-121-22 Supplier Invitrogen™ Supplier No. 13599785
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The MECA-367 monoclonal antibody reacts with mouse mucosal vascular addressin-1 (MAdCAM-1), a 50-60 kDa member of the Ig superfamily. This antigen is expressed in mucosal lymphoid tissue and lamina propria and is a counter-receptor for CD62L and CD49d. MAdCAM-1 is involved in homing of subpopulation of memory T cells to the gut lamina propria, and recirculation of naive lymphocytes in Peyer's patches and mesenteric lymph nodes. Applications Reported: MECA-367 can be used in flow cytometric analysis, immunohistochemical staining of frozen mouse tissue sections, blocking in adhesion assays, immunoprecipitation and immunoblotting (WB). Applications Tested: The MECA-367 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes and can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

The protein encoded by this gene is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAdCAM1
Applications Flow Cytometry
Classification Monoclonal
Clone MECA-367
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene MADCAM1
Gene Accession No. Q61826
Gene Alias AV211525; hMAdCAM-1; MACAM1; MADCAM1; MAdCAM-1; mMAdCAM-1; mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1; mucosal vascular addressin cell adhesion molecule 1; rMAdCAM-1
Gene Symbols MADCAM1
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17123
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.