Learn More
MADCAM1 Mouse anti-Human, Clone: OTI1D6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506920

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MAdCAM-1 protein is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.Specifications
MADCAM1 | |
Monoclonal | |
Unconjugated | |
MADCAM1 | |
MACAM1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8174 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1D6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13477 | |
MADCAM1 | |
Full length human recombit protein of human MADCAM1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.