Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAdCAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109929 Supplier Invitrogen™ Supplier No. PA5109929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRTFQHTVQL LVYAFPDQLT VSPAALVPGD PEVACTAHKV TPVDPNALSF SLLVGGQELE GAQALGPEVQ EEEEEPQGDE DVLFRVTERW RLPPLGTPVP P.

The protein encoded by this gene is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAdCAM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MADCAM1
Gene Accession No. Q13477
Gene Alias AV211525; hMAdCAM-1; MACAM1; MADCAM1; MAdCAM-1; mMAdCAM-1; mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1; mucosal vascular addressin cell adhesion molecule 1; rMAdCAM-1
Gene Symbols MADCAM1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human MADCAM1 (NP_570116.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.