Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MADD Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306250AF700
Description
MADD Polyclonal specifically detects MADD in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
MADD | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
DENNFLJ36300, Differentially expressed in normal and neoplastic cells, FLJ35600, IG20, Insulinoma glucagonoma clone 20, insulinoma-glucagonoma protein 20, KIAA0358RAB3GEP, MAP kinase-activating death domain protein, MAP-kinase activating death domain, Rab3 GDP/GTP exchange factor | |
Produced in rabbits immunized with E. coli-derived Human MADD fragment. | |
0.1 mL | |
Signal Transduction | |
8567 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction