Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MADD Antibody (SR1358), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32255050UL
Description
MADD Monoclonal antibody specifically detects MADD in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
MADD | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1358 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
DENNFLJ36300, Differentially expressed in normal and neoplastic cells, FLJ35600, IG20, Insulinoma glucagonoma clone 20, insulinoma-glucagonoma protein 20, KIAA0358RAB3GEP, MAP kinase-activating death domain protein, MAP-kinase activating death domain, Rab3 GDP/GTP exchange factor | |
A synthesized peptide derived from human MADD (Uniprot #: Q8WXG6) | |
50 μg | |
Signal Transduction | |
8567 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction