Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAEL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585064 Supplier Invitrogen™ Supplier No. PA585064
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVFTPLRRPG MLVPKQNVSP PDMSALSLKG DQALLGGIFY FLNIFSHGEL PPHCEQRFLP CEIGCVKYSL QEGIMADFH Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 90%.

The mammalian homolog of the Drosophila protein Maelstrom is expressed in the male germline and localizes to the sex body in spermatocytes and the chromatid body in round spermatids. Similar to its expression in Drosophila, Maelstrom is a component of nuages, a germ-cell specific organelle and is thought to be essential for spermatogenesis and transposon repression during meiosis. In humans, Maelstrom has been found to be expressed only in the testis and in various cancer cell lines. Treatment of these cell lines with the demethylating agent 5'-Aza-2-Deoxycytidine significantly upregulated Maelstrom levels, indicating that its expression is regulated by DNA methylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAEL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAEL
Gene Accession No. Q96JY0
Gene Alias 4933405K18Rik; AU019877; cancer/testis antigen 128; CT128; MAEL; maelstrom homolog; maelstrom homolog (Drosophila); maelstrom spermatogenic transposon silencer; protein maelstrom homolog; RGD1309333; RP11-102C16.1; SPATA35; spermatogenesis associated 35; testicular tissue protein Li 116
Gene Symbols MAEL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAEL. Recombinant protein control fragment (Product #RP-94271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84944
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.