Learn More
Mopec Maestro Encore Safety Sash
Enhance laboratory safety and workflow with the AX124 Safety Sash.
Supplier: Mopec AX124R048
Description
- Designed to reduce exposure to formalin fumes and hazardous chemical spills and splashes
- Engineered to work with PathCam imaging systems, Ensemble accessories, and formalin dispensing
- Frameless door
- Friction hinges
- 304 stainless steel sheetmetal and hardware
- Ideal for open formalin processing operations
- Ensures maximum fume capture even in high traffic cross flow laboratory conditions
Specifications
Grossing Station | |
Stainless Steel (304) | |
Safety Sash, Right | |
48 Units |