Learn More
Mopec Maestro Encore Safety Sash
SDP
Enhance laboratory safety and workflow with the AX124 Safety Sash.
$2352.04
Specifications
Product Type Grossing Station
Material Stainless Steel (304)
View More Specs
Products 6
Catalog Number Mfr. No. For Use With (Equipment) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Quantity Price Quantity & Availability  
23-987-032
SDP
View Documents
Mopec
AX124L048
Safety Sash, Left 48 Units
Each for $2,352.04
Only null left
Add to Cart
23-987-033
SDP
View Documents
Mopec
AX124L060
Safety Sash, Left 60 Units
Each for $2,352.04
Only null left
Add to Cart
23-987-034
SDP
View Documents
Mopec
AX124L072
Safety Sash, Left 72 Units
Each for $2,352.04
Only null left
Add to Cart
23-987-035
SDP
View Documents
Mopec
AX124R048
Safety Sash, Right 48 Units
Each for $2,352.04
Only null left
Add to Cart
23-987-036
SDP
View Documents
Mopec
AX124R060
Safety Sash, Right 60 Units
Each for $2,352.04
Only null left
Add to Cart
23-987-037
SDP
View Documents
Mopec
AX124R072
Safety Sash, Right 72 Units
Each for $2,352.04
Only null left
Add to Cart
Description
Description
  • Designed to reduce exposure to formalin fumes and hazardous chemical spills and splashes
  • Engineered to work with PathCam imaging systems, Ensemble accessories, and formalin dispensing
  • Frameless door
  • Friction hinges
  • 304 stainless steel sheetmetal and hardware
  • Ideal for open formalin processing operations
  • Ensures maximum fume capture even in high traffic cross flow laboratory conditions
Specifications
Specifications
Grossing Station
Stainless Steel (304)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.