Learn More
Mopec Maestro Exhaust Dampening Kit
Designed to integrate adjustable exhaust dampeners into your grossing station to help your HVAC/facilities
Supplier: Mopec AX111072
Description
- Designed to integrate adjustable exhaust dampeners into your grossing station to help your HVAC/facilities personnel balance the airflow to the grossing stations
- 304 stainless steel sheetmetal and hardware
- Fits the Mopec Maestro and Maestro Encore grossing stations of the accompanying sized unit
Specifications
Grossing Station | |
72 in. |