Learn More
Mopec Maestro Exhaust Dampening Kit
Designed to integrate adjustable exhaust dampeners into your grossing station to help your HVAC/facilities
$972.91
Catalog Number | Mfr. No. | Length (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Price | Quantity & Availability | |||||
23-987-024
![]() |
Mopec
AX111048 |
48 in. |
Each for $972.91
|
|
|||||
23-987-025
![]() |
Mopec
AX111060 |
60 in. |
Each for $972.91
|
|
|||||
23-987-026
![]() |
Mopec
AX111072 |
72 in. |
Each for $972.91
|
|
|||||
23-987-027
![]() |
Mopec
AX111096 |
96 in. |
Each for $972.91
|
|
|||||
Description
- Designed to integrate adjustable exhaust dampeners into your grossing station to help your HVAC/facilities personnel balance the airflow to the grossing stations
- 304 stainless steel sheetmetal and hardware
- Fits the Mopec Maestro and Maestro Encore grossing stations of the accompanying sized unit
Specifications
Grossing Station |