Learn More
Mopec Maestro Exhaust Dampening Kit
SDP
Designed to integrate adjustable exhaust dampeners into your grossing station to help your HVAC/facilities
Supplier:  Mopec AX111096
 View more versions of this product
Catalog No. 23-987-027
Only null left
Add to Cart
Description
Description
  • Designed to integrate adjustable exhaust dampeners into your grossing station to help your HVAC/facilities personnel balance the airflow to the grossing stations
  • 304 stainless steel sheetmetal and hardware
  • Fits the Mopec Maestro and Maestro Encore grossing stations of the accompanying sized unit
Specifications
Specifications
Grossing Station
96 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.