Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mopec Maestro Non-Elevating Leg Frame

Includes leg frame and leveling feet with 1.625 in. of adjustment
Supplier: Mopec MB1030072
Description
- Designed for customers who do not need the elevation function on a standard Elevating Maestro or Maestro Encore grossing station
- Includes leg frame and leveling feet with 1.625 in. of adjustment
- 304 Stainless steel, 180 grit finish
- 11 ga 304 stainless steel seismic anchor brackets included, with 3/8 in. 304 stainless hardware
- Compatible with all Maestro and Maestro Encore Non-Elevating grossing stations
Specifications
Grossing Station | |
29 in. | |
72 in. |
Stainless Steel | |
32 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction