Learn More
Mopec Maestro Recirculating to In-House Facility Vented Conversion Kit
SDP
Designed to convert recirculating grossing stations into in-house ventilation
Supplier:  Mopec AX110072
 View more versions of this product
Catalog No. 23-987-022
Only null left
Add to Cart
Description
Description
  • Designed to convert recirculating grossing stations into in-house ventilation
  • Bolt-on panel with 8 in duct stubs which allows the grossing station to be connected to the facility ventilation system
  • Includes panel, duct tubing, and clamps
  • 304 stainless steel sheetmetal and hardware
  • Fits the Mopec Maestro and Maestro Encore grossing stations of the accompanying sized unit
Specifications
Specifications
Grossing Station
Stainless Steel (304)
72 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.