Learn More
Mopec Maestro Recirculating to In-House Facility Vented Conversion Kit
SDP
Designed to convert recirculating grossing stations into in-house ventilation
$1743.11 - $1909.40
Specifications
Product Type Grossing Station
Material Stainless Steel (304)
View More Specs
Products 4
Catalog Number Mfr. No. Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Price Quantity & Availability  
23-987-020
SDP
View Documents
Mopec
AX110048
48 in.
Each for $1,743.11
Only null left
Add to Cart
23-987-021
SDP
View Documents
Mopec
AX110060
60 in.
Each for $1,743.11
Only null left
Add to Cart
23-987-022
SDP
View Documents
Mopec
AX110072
72 in.
Each for $1,743.11
Only null left
Add to Cart
23-987-023
SDP
View Documents
Mopec
AX110096
96 in.
Each for $1,909.40
Only null left
Add to Cart
Description
Description
  • Designed to convert recirculating grossing stations into in-house ventilation
  • Bolt-on panel with 8 in duct stubs which allows the grossing station to be connected to the facility ventilation system
  • Includes panel, duct tubing, and clamps
  • 304 stainless steel sheetmetal and hardware
  • Fits the Mopec Maestro and Maestro Encore grossing stations of the accompanying sized unit
Specifications
Specifications
Grossing Station
Stainless Steel (304)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.