Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555195 Supplier Invitrogen™ Supplier No. PA555195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKLLENSSFE AINSQLTVET GDAHIIGRIE SYSCKMAGDD KHMFKQFCQE GQPHVLEALS PPQTSGLS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a protein that is similar to Maf1, a Saccharomyces cerevisiae protein highly conserved in eukaryotic cells. Yeast Maf1 is a negative effector of RNA polymerase III (Pol III). It responds to changes in the cellular environment and represses pol III transcription. Biochemical studies identified the initiation factor TFIIIB as a target for Maf1-dependent repression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAF1
Gene Accession No. Q9H063
Gene Alias 1110068E11Rik; AU042856; homolog of yeast MAF1; MAF1; MAF1 homolog (S. cerevisiae); MAF1 homolog, negative regulator of RNA polymerase III; MAF1 negative regulator of RNA polymerase III; repressor of RNA polymerase III transcription MAF1 homolog
Gene Symbols MAF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAF1. Recombinant protein control fragment (Product #RP-93931).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84232
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.