Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MafA Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $426.00

Specifications

Antigen MafA
Concentration 0.5 mg/mL
Dilution Western Blot 0.25-0.5ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 24509877), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24509877), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24509877), ICC/IF (Negative), Chromatin Immunoprecipitation (ChIP)
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224636SS
SDP
View Documents
Novus Biologicals
NBP224636SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224636
SDP
View Documents
Novus Biologicals
NBP224636
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

MafA Polyclonal specifically detects MafA in Human, Mouse, Rat, Opossum samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, ICC/IF (Negative), Chromatin Immunoprecipitation (ChIP).
Specifications

Specifications

MafA
Western Blot 0.25-0.5ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 24509877), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24509877), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24509877), ICC/IF (Negative), Chromatin Immunoprecipitation (ChIP)
Polyclonal
Purified
RUO
Human, Mouse, Rat, Opossum
Q8NHW3
389692
A portion of amino acids 300-350 of human MafA was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
hMafA, Pancreatic beta-cell-specific transcriptional activator, RIPE3B1, transcription factor MafA, Transcription factor RIPE3b1, V-maf musculoaponeurotic fibrosarcoma oncogene homolog A, v-maf musculoaponeurotic fibrosarcoma oncogene homolog A (avian)
MAFA
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, pig and bovine, 87% homologous in pufferfish and 81% homologous in zebrafish.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.