Learn More
MafA Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300611AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Human
MAFA is a transcription factor that binds RIPE3b, a conserved enhancer element that regulates pancreatic beta cell-specific expression of the insulin gene. MAFA cooperates synergistically with NEUROD1 and PDX1. Phosphorylation by GSK3 increases its transcriptional activity and is required for its ocogenic activity. Serves as an ocogene or tumor supressor depending on the cell context.Specifications
MafA | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8CF90 | |
Mafa | |
Between 300 and C-term | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Mafa | |
RIPE3b1, hMafA, pancreatic beta-cell-specific transcriptional activator, transcription factor MafA, transcription factor RIPE3b1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
378435 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.