Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAFB Monoclonal Antibody (OTI1E9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA800290

Description
The protein encoded by this gene is a basic leucine zipper transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns.Specifications
MAFB | |
Monoclonal | |
Unconjugated | |
MAFB | |
DURS3, KRML, MCTO | |
Mouse | |
Affinity Chromatography | |
RUO | |
9935 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1E9 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y5Q3 | |
MAFB | |
Human recombit protein fragment corresponding to amino acids 32-320 of human MAFB produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction