Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAFB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540757 Supplier Invitrogen™ Supplier No. PA540757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RPGRPCTRLQ PAGSVSSTPL STPCSSVPSS PSFSPTEQKT HLEDLYWMAS Sequence homology: Cow: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

The protein encoded by this gene is a basic leucine zipper transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAFB
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAFB
Gene Accession No. P54842, Q9Y5Q3
Gene Alias b-maf; c-Maf; kr; Kreisler; Kreisler (maf-related) leucine zipper protein; Kreisler maf-related leucine zipper homolog; Kreisler maf-related leucine zipper homolog (b-maf); KRML; Krml1; LOW QUALITY PROTEIN: transcription factor MafB; maf; MAF bZIP transcription factor B; Maf1; MAFB; maf-B; MAFB/Kreisler basic region/leucine zipper transcription factor; MCTO; MGC43127; segmentation protein Kr; si:ch73-337g9.1; transcription factor Maf; Transcription factor Maf-1; transcription factor MafB; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog B; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog b (paralog b); v-maf musculoaponeurotic fibrosarcoma oncogene family, protein B; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein B (avian); V-maf musculoaponeurotic fibrosarcoma oncogene homolog B
Gene Symbols MAFB
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MAFB (aa 41-90).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54264, 9935
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.