Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAFF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565947 Supplier Invitrogen™ Supplier No. PA565947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSSKALKIKRE LSENTPHLSD EALMGLSVRE L Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

The protein encoded by this gene is a basic leucine zipper (bZIP) transcription factor that lacks a transactivation domain. It is known to bind the US-2 DNA element in the promoter of the oxytocin receptor (OTR) gene and most likely heterodimerizes with other leucine zipper-containing proteins to enhance expression of the OTR gene during term pregnancy. The encoded protein can also form homodimers, and since it lacks a transactivation domain, the homodimer may act as a repressor of transcription. This gene may also be involved in the cellular stress response. Multiple transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAFF
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Maff
Gene Accession No. Q9ULX9
Gene Alias hMafF; hypothetical protein MGC63868; LOW QUALITY PROTEIN: transcription factor MafF; MAF bZIP transcription factor F; Maff; Transcription factor MafF; transcription factor mafT; U-Maf; v-maf avian musculoaponeurotic fibrosarcoma oncogene family protein F; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein F; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein F (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog f; zgc:63868
Gene Symbols Maff
Host Species Rabbit
Immunogen Recombinant Human MAFF. Recombinant protein control fragment (Product #RP-106628).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23764
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.