Learn More
Invitrogen™ MAFG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590907
Description
Immunogen sequence: MTTPNKGNKA LKVKREPGEN GTSLTDEELV TMSVRELNQH LRGLSKEEIV QLKQRRRTLK NRGYAASCRV KRVTQKEELE KQKAELQQEV EKLASENASM KLELDALRSK YEALQTFART VARSPVAPAR GPLAAGLGPL VPGKVAATSV ITIVKSKTDA RS; Positive Samples: U-87MG, A-549, HT-29, 293T; Cellular Location: Nucleus.
Globin gene expression is regulated through nuclear factor erythroid-2 (NFE2) elements located in enhancer-like locus control regions positioned many kb upstream of alpha- and beta-gene clusters (summarized by Blank et al., 1997 [PubMed 9166829]). NFE2 DNA-binding activity consists of a heterodimer containing a ubiquitous small Maf protein (MafF, MIM 604877; MafG; or MafK, MIM 600197) and the tissue-restricted protein p45 NFE2 (MIM 601490). Both subunits are members of the activator protein-1-like superfamily of basic leucine zipper (bZIP) proteins (see MIM 165160).
Specifications
MAFG | |
Polyclonal | |
Unconjugated | |
MAFG | |
AA545192; avian musculoaponeurotic fibrosarcoma virus (v-maf) AS42 oncogene, protein G; basic leucine zipper transcription factor MafG; C630022N07Rik; hMAF; MAF bZIP transcription factor G; MAFG; MGC13090; MGC20149; Transcription factor MafG; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog G | |
Rabbit | |
Affinity Chromatography | |
RUO | |
17134, 4097 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O15525, O54790 | |
MAFG | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-162 of human MAFG (NP_0023501). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.