Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAFG Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5107166

Catalog No. PIPA5107166


Only null left
Add to Cart

Description

Description

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 93%.

Globin gene expression is regulated through nuclear factor erythroid-2 (NFE2) elements located in enhancer-like locus control regions positioned many kb upstream of alpha- and beta-gene clusters (summarized by Blank et al., 1997 [PubMed 9166829]). NFE2 DNA-binding activity consists of a heterodimer containing a ubiquitous small Maf protein (MafF, MIM 604877; MafG; or MafK, MIM 600197) and the tissue-restricted protein p45 NFE2 (MIM 601490). Both subunits are members of the activator protein-1-like superfamily of basic leucine zipper (bZIP) proteins (see MIM 165160).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAFG
Polyclonal
Unconjugated
MAFG
AA545192; avian musculoaponeurotic fibrosarcoma virus (v-maf) AS42 oncogene, protein G; basic leucine zipper transcription factor MafG; C630022N07Rik; hMAF; MAF bZIP transcription factor G; MAFG; MGC13090; MGC20149; Transcription factor MafG; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog G
Rabbit
Affinity Chromatography
RUO
4097, 64188
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
O15525, Q76MX4
MAFG
Synthetic peptide directed towards the N terminal region of human MAFG.
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.