Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MafG Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | MafG |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MafG Polyclonal specifically detects MafG in Human samples. It is validated for Western Blot.Specifications
MafG | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
4097 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
basic leucine zipper transcription factor MafG, hMAF, MGC13090, MGC20149, transcription factor MafG, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein G, V-maf musculoaponeurotic fibrosarcoma oncogene homolog G, v-maf musculoaponeurotic fibrosarcoma oncogene homolog G (avian) | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-162 of human MAFG (NP_002350.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title