Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAFK Monoclonal Antibody (1328CT786.105.125)
GREENER_CHOICE

Catalog No. PIMA524732
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524732 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524732 Supplier Invitrogen™ Supplier No. MA524732
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAFK Monoclonal Antibody for Flow, Western Blot

Since they lack a putative transactivation domain, the small Mafs behave as transcriptional repressors when they dimerize among themselves. However, they seem to serve as transcriptional activators by dimerizing with other (usually larger) basic-zipper proteins and recruiting them to specific DNA-binding sites. Small Maf proteins heterodimerize with Fos and may act as competitive repressors of the NF-E2 transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAFK
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1328CT786.105.125
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MAFK
Gene Accession No. O60675
Gene Alias AW061068; basic-leucine zipper transcription factor MafK; erythroid transcription factor NF-E2 p18 subunit; FLJ32205; MAF bZIP transcription factor K; Mafk; MGC71717; NF-E2; Nfe2u; nuclear factor erythroid-2, ubiquitous (p18); nuclear factor, erythroid derived 2, ubiquitous; P18; p18 NF-E2; transcription factor MafK; v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein K (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog K
Gene Symbols MAFK
Host Species Mouse
Immunogen KLH conjugated synthetic peptide of human MAFK.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.