Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAFK Monoclonal Antibody (381923)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524120

Catalog No. PIMA524120


Only null left
Add to Cart

Description

Description

In Western blots, no cross-reactivity with recombinant human (rh) MafB or rhMafF is observed. In Western blots, prolonged film exposures may slightly detect rhMafG. Reconstitute at 0.5 mg/mL in sterile PBS.

Since they lack a putative transactivation domain, the small Mafs behave as transcriptional repressors when they dimerize among themselves. However, they seem to serve as transcriptional activators by dimerizing with other (usually larger) basic-zipper proteins and recruiting them to specific DNA-binding sites. Small Maf proteins heterodimerize with Fos and may act as competitive repressors of the NF-E2 transcription factor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAFK
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
O60675, Q61827
MAFK
E. coli-derived recombinant human MafK Thr2-Ser156.
100 μg
Primary
Human, Mouse
Antibody
IgG2b
Western Blot
381923
Unconjugated
MAFK
AW061068; basic-leucine zipper transcription factor MafK; erythroid transcription factor NF-E2 p18 subunit; FLJ32205; MAF bZIP transcription factor K; Mafk; MGC71717; NF-E2; Nfe2u; nuclear factor erythroid-2, ubiquitous (p18); nuclear factor, erythroid derived 2, ubiquitous; P18; p18 NF-E2; transcription factor MafK; v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein K (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog K
Mouse
Protein A/G
RUO
17135, 7975
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.