Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAFK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115534 Supplier Invitrogen™ Supplier No. PA5115534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAFK.

Since they lack a putative transactivation domain, the small Mafs behave as transcriptional repressors when they dimerize among themselves. However, they seem to serve as transcriptional activators by dimerizing with other (usually larger) basic-zipper proteins and recruiting them to specific DNA-binding sites. Small Maf proteins heterodimerize with Fos and may act as competitive repressors of the NF-E2 transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAFK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MAFK
Gene Accession No. O60675, Q61827
Gene Alias AW061068; basic-leucine zipper transcription factor MafK; erythroid transcription factor NF-E2 p18 subunit; FLJ32205; MAF bZIP transcription factor K; Mafk; MGC71717; NF-E2; Nfe2u; nuclear factor erythroid-2, ubiquitous (p18); nuclear factor, erythroid derived 2, ubiquitous; P18; p18 NF-E2; transcription factor MafK; v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein K (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog K
Gene Symbols MAFK
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAFK(Accession O60675), corresponding to amino acid residues M1-K46.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17135, 246760, 7975
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.