Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAFK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566349
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566349 Supplier Invitrogen™ Supplier No. PA566349
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITIVKSTELSS TSVPFSAAS Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

Since they lack a putative transactivation domain, the small Mafs behave as transcriptional repressors when they dimerize among themselves. However, they seem to serve as transcriptional activators by dimerizing with other (usually larger) basic-zipper proteins and recruiting them to specific DNA-binding sites. Small Maf proteins heterodimerize with Fos and may act as competitive repressors of the NF-E2 transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAFK
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAFK
Gene Accession No. O60675
Gene Alias AW061068; basic-leucine zipper transcription factor MafK; erythroid transcription factor NF-E2 p18 subunit; FLJ32205; MAF bZIP transcription factor K; Mafk; MGC71717; NF-E2; Nfe2u; nuclear factor erythroid-2, ubiquitous (p18); nuclear factor, erythroid derived 2, ubiquitous; P18; p18 NF-E2; transcription factor MafK; v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein K (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog K
Gene Symbols MAFK
Host Species Rabbit
Immunogen Recombinant Human MAFK. Recombinant protein control fragment (Product #RP-107032).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.