Learn More
Invitrogen™ MAG Polyclonal Antibody

Description
MAG Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | MAG |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | MAG |
| Gene Accession No. | P07722, P20916, P20917 |
| Gene Alias | 1B236; Brain neuron cytoplasmic protein 3; Gma; MAG; myelin associated glycoprotein; myelin-associated glycoprotein; sialic acid binding Ig-like lectin 4A; sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; SIGLEC4A; siglec-4a; S-MAG; S-MAG (C-term.); SPG75 |
| Gene Symbols | MAG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.