Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAG Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0182 Supplier Bioss Supplier No. BS0258R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.

Specifications

Antigen MAG
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAG
Gene Accession No. P20916
Gene Alias 1B236; Brain neuron cytoplasmic protein 3; GMA; MAG; myelin associated glycoprotein; Myelin-associated glycoprotein; sialic acid binding Ig-like lectin 4A; Sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; SIGLEC4A; Siglec-4a; S-MAG; S-MAG (C-term.); SPG75
Gene Symbols MAG
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAG.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4099
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.