Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Magbeads FastDNA/RNA Kit for Virus

Catalog No. MP116571192 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
:
192 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP116571192 192 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116571192 Supplier MP Biomedicals™ Supplier No. 116571192
Only null left
Add to Cart
Add to Cart

MagBeads FastDNA/RNA Kit for Virus is intended for extracting total viral nucleic acid from cell-free/low-cell content biological samples such as body fluids, serum, plasma, soaking solution, tissue homogenate, and culture supernatant.

MagBeads FastDNA/RNA Kit for Virus is based on the purification method of high binding magnetic particles:

  • The sample is lysed and digested with Protease
  • DNA/RNA is released into the lysate
  • After addition of magnetic particles and binding solution, DNA/RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption
  • The adsorbed particles are washed with washing solution to remove proteins and impurities, washed with ethanol to remove salts, and finally DNA/RNA is eluted with Nuclease Free Water
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
Sample Type Virus
Sequencing Type DNA and RNA Sequencing
No. of Reactions 192 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.