Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MagBeads FastRNA™ Kit for Virus, 50 preps

Catalog No. MP116578050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116578050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116578050 Supplier MP Biomedicals™ Supplier No. 116578050
Only null left
Add to Cart
Add to Cart

The MagBeads FastRNA™ Kit for Virus is designed to quickly and effectively isolate high-quality total viral RNA from cell culture media, swabs, and bodily fluids such as blood, plasma, serum, saliva, and more in 60 minutes.

  • High-quality viral RNA extraction from a variety of samples
  • Compatible with cell culture media, swabs, and bodily fluids
  • Magnetic bead-based method designed for manual and automated workflows
  • Flexible mid- to high-throughput processing
  • No phenol chloroform extractions or toxic reagents
  • Less than 60 minutes to process samples
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store Carrier RNA and Proteinase K at 2 to 8 °C. Store other kit components at room temperature, Dry environment.
Sample Type Cell Culture Media, Swabs, and Bodily Fluids
Includes 1.2 mL Magnetic Beads VRM, 32 mL Lysis Buffer VRM, 16.5 mL Wash Buffer VRM 1, 6 mL Wash Buffer VRM 2, 10 mL RNase-free Water, 1 vial Carrier RNA, 1 vial Proteinase K, 1.5 mL Proteinase K Buffer, 2 x 96-deep well plates
Product Type RNA Extraction Kit
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.