Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Magbeads FastRNA Kit
Shop All MP Biomedicals, Inc Products
Click to view available options
:
96 Preps
Description
- Intended for simple and rapid extraction of total RNA from tissue and culture cells samples
- Based on superparamagnetic particles purification technology, no phenol-chloroform extraction or alcohol precipitation
- The whole extraction process is only 60 minutes. Purified RNA is ready for downstream applications such as RTPCR, virus RNA testing and so on
- The kits buffers can be used for both manual extraction process, automatic nucleic acid extraction machines

Specifications
Specifications
Format | Kit |
Isolation Technology | Magnetic Bead |
Scale | Mini |
Sample Type | Tissue, Cell Cultures |
Sequencing Type | DNA and RNA Sequencing |
No. of Reactions | 96 Preps |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction