Learn More
R&D Systems™ MagCellect Human NK Cell Isolation Kit
SDP
Catalog No. MAGH109 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
MAGH109 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. MAGH109 Supplier R&D Systems™ Supplier No. MAGH109
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Designed to isolate Natural Killer cells via a negative selection principle
Typical purity of recovered NK cells ranges between 80% and 90%
MagCellect Streptavidin Ferrofluid is added to the reaction which allows the streptavidin-coated nanoparticles to interact with biotinylated antibody tagged cells, the unwanted cell fraction
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit