Learn More
Description
CD8 is a heterodimeric glycoprotein consisting of an alpha and beta chain. It is expressed on cytolytic T cells and functions in conjunction with the T cell receptor in the recognition of MHC/peptide complexes.
Typical recoveries range between 5% and 35% and the purity of recovered cells ranges between 80 - 90%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
These Ferrofluids have a diameter of 150nm and as a result behave like colloidal particles, offering a higher ligand capacity per mass compared to other, larger magnetic particles
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 1 Kit |
| Type | Cell Isolation Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
