Learn More
Description
Epithelial Cellular Adhesion Molecule (EpCAM), also known as KS1/4, gp40, GA733-2, 17-1A, and TROP-1, is a transmembrane glycoprotein that functions as a homophilic Ca2+-independent adhesion molecule. EpCAM is expressed in fetal lung, kidney, liver, pancreas, skin, and germ cells, and in adult epithelia. EpCAM expression is elevated in human malignant neoplasia, with most carcinomas expressing EpCAM.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Detection Method | ELISA |
| Product Line | R&D Systems |
| Quantity | 1 Kit |
| Type | Cell Isolation Kit |
For Research Use Only. Not for use in Humans.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
