Learn More
R&D Systems™ MagCellect Rat CD3+ T Cell Isolation Kit
SDP
Catalog No. MAGR301B Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
MAGR301B 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. MAGR301B Supplier R&D Systems™ Supplier No. MAGR301B
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
The CD3 complex functions to transduce intracellular signals during TCR antigen recognition
  • The T cell receptor (TCR)/CD3 complex consists of two variable antigen-recognition receptor chains (TCR-alpha/TCR-beta or TCR-gamma/TCR-delta) that are non-covalently linked to at least four different invariant chains: CD3 gamma, CD3 delta, CD3 epsilon, and the CD3 zeta chain that exists as either a homodimer or as a heterodimer with its splice-variant the CD3 eta chain
  • The CD3 complex functions to transduce intracellular signals during TCR antigen recognition
  • CD3 is expressed early in thymocyte development, and on a subset of NK cells
  • CD3 epsilon is one of at least three invariant proteins that associate with the variable antigen recognition chains of the T cell receptor and function in signal transduction
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit