Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ MAGE-1 Ab-6, Mouse Monoclonal Antibody

Catalog No. MS1836S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1836S 1 mL Concentrated
MS1836S0 100 μL Concentrated
MS1836S1 500 μL Concentrated
MS1836R1 1 mL Prediluted
MS1836R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1836S Supplier Epredia™ Supplier No. MS1836S

Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ MAGE-1 Ab-6, Mouse Monoclonal Antibody provides accurate, reproducible results.

Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor-directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). An important group of antigens is coded by the MAGE family of genes. It was identified that melanomas and primary glial brain tumors express common melanoma associated antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various histological types (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy

Host Species: Mouse

Clone: 6C1

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Full-length recombinant human MAGE-1 protein

Molecular Weight: 42-46kDa

Positive Control: Hepatocellular CA, normal testis

Cellular Localization: Cytoplasmic, occasionally nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MAGE-1 Ab-6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6C1
Conjugate Unconjugated
Host Species Mouse
Immunogen Full length recombinant human MAGE-1 protein
Purification Method Hepatocellular CA, normal testis
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.